Search
Search

SULTR4;2| Gene Symbol: SULTR4;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202942.1 | NP_001189871.1 | 5 - 6 | 695 | 89 | |
| NM_001337994.1 | NP_001327651.1 | 4 - 5 | 882 | 89 | ||
| NM_112087.3 | NP_187858.1 | 4 - 5 | 957 | 89 | ||
| GenBank mRNA | AB052775.1 | - | 4 - 5 | 740 | 89 | |
| AK117615.1 | - | 5 - 6 | 701 | 89 | ||
Set Membership: |
|