Search
Search

AT4G28706| Gene Symbol: AT4G28706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119014.4 | NP_567816.1 | 1 - 2 | 124 | 80 | |
| GenBank mRNA | AF428378.1 | - | 1 - 2 | 124 | 80 | |
| AY125545.1 | - | 1 - 2 | 74 | 80 | ||
Set Membership: |
|