Search
Search

PAP2| Gene Symbol: PAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119052.4 | NP_194637.1 | 2 - 3 | 826 | 157 | |
| GenBank mRNA | AF087936.1 | - | 2 - 3 | 678 | 157 | |
| AF332402.1 | - | 2 - 3 | 678 | 157 | ||
| AJ458326.1 | - | 2 - 3 | 678 | 157 | ||
| AY053404.1 | - | 2 - 3 | 683 | 157 | ||
| AY133516.1 | - | 2 - 3 | 678 | 157 | ||
Set Membership: |
|