Search
Search

ATMND1| Gene Symbol: ATMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085000.2 | NP_001078469.1 | 7 - 8 | 707 | 73 | |
| NM_119061.4 | NP_194646.2 | 7 - 8 | 612 | 73 | ||
| GenBank mRNA | AK117713.1 | - | 7 - 8 | 561 | 73 | |
| AM162278.1 | - | 6 - 7 | 463 | 73 | ||
| BT005435.1 | - | 6 - 7 | 463 | 73 | ||
| DQ248000.1 | - | 7 - 8 | 584 | 73 | ||
Set Membership: |
|