Search
Search

CKX4| Gene Symbol: CKX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341977.1 | NP_001328306.1 | 2 - 3 | 921 | 67 | |
| NM_119120.2 | NP_194703.1 | 2 - 3 | 921 | 67 | ||
| NM_179139.1 | NP_849470.1 | 2 - 3 | 839 | 67 | ||
| GenBank mRNA | AY054460.1 | - | 2 - 3 | 841 | 67 | |
| BT000179.1 | - | 2 - 3 | 753 | 67 | ||
Set Membership: |
|