Search
Search

CFIM-25| Gene Symbol: CFIM-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119128.3 | NP_567835.1 | 3 - 4 | 516 | 84 | |
| GenBank mRNA | AY048229.1 | - | 3 - 4 | 479 | 84 | |
| AY085984.1 | - | 3 - 4 | 481 | 84 | ||
| AY091704.1 | - | 3 - 4 | 317 | 84 | ||
Set Membership: |
|