Search
Search

HA2| Gene Symbol: HA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203941.2 | NP_001190870.1 | 12 - 13 | 2622 | 63 | |
| NM_119165.4 | NP_194748.1 | 13 - 14 | 2712 | 63 | ||
| GenBank mRNA | AY035075.1 | - | 13 - 14 | 2513 | 63 | |
| BT000781.1 | - | 13 - 14 | 2513 | 63 | ||
| BT001969.1 | - | 13 - 14 | 2385 | 63 | ||
Set Membership: |
|