Search
Search

HHP2| Gene Symbol: HHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119232.3 | NP_194814.2 | 2 - 3 | 555 | 102 | |
| NM_202914.1 | NP_974643.1 | 2 - 3 | 495 | 102 | ||
| GenBank mRNA | AY267331.1 | - | 2 - 3 | 328 | 102 | |
| BT021124.1 | - | 2 - 3 | 427 | 102 | ||
| BT025258.1 | - | 2 - 3 | 328 | 102 | ||
Set Membership: |
|