Search
Search

NFD1| Gene Symbol: NFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119240.4 | NP_567861.1 | 2 - 3 | 638 | 81 | |
| GenBank mRNA | AB493710.1 | - | 2 - 3 | 517 | 81 | |
| AK228212.1 | - | 2 - 3 | 547 | 81 | ||
| AY088563.1 | - | 2 - 3 | 552 | 81 | ||
| BT005870.1 | - | 2 - 3 | 517 | 81 | ||
Set Membership: |
|