Search
Search

TOP1ALPHA| Gene Symbol: TOP1ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203614.1 | NP_001190543.1 | 14 - 15 | 2654 | 91 | |
| NM_001203615.1 | NP_001190544.1 | 8 - 9 | 1591 | 91 | ||
| NM_124912.5 | NP_200341.1 | 14 - 15 | 2666 | 91 | ||
| GenBank mRNA | X57544.1 | - | 2591 | 91 | ||
Set Membership: |
|