Search
Search

SVL1| Gene Symbol: SVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124930.4 | NP_200359.1 | 4 - 5 | 1179 | 126 | |
| GenBank mRNA | BT015819.1 | - | 4 - 5 | 1091 | 126 | |
| BT020207.1 | - | 4 - 5 | 1044 | 126 | ||
Set Membership: |
|