Search
Search

SVL1| Gene Symbol: SVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124930.4 | NP_200359.1 | 8 - 9 | 2141 | 91 | |
| GenBank mRNA | BT015819.1 | - | 8 - 9 | 2053 | 91 | |
| BT020207.1 | - | 8 - 9 | 2006 | 91 | ||
Set Membership: |
|