Search
Search

TAFII15| Gene Symbol: TAFII15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342109.1 | NP_001329548.1 | 2 - 3 | 482 | 86 | |
| NM_119321.4 | NP_194900.1 | 2 - 3 | 373 | 86 | ||
| NM_179150.4 | NP_849481.1 | 2 - 3 | 356 | 86 | ||
| GenBank mRNA | AF344879.1 | - | 2 - 3 | 283 | 86 | |
| AF352813.1 | - | 2 - 3 | 283 | 86 | ||
| AK317292.1 | - | 2 - 3 | 288 | 86 | ||
| AY086560.1 | - | 2 - 3 | 288 | 86 | ||
| AY463628.1 | - | 1 - 2 | 96 | 86 | ||
| Y12904.1 | - | 273 | 86 | |||
Set Membership: |
|