Search
Search

MOM| Gene Symbol: MOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331755.1 | NP_001322007.1 | 4 - 5 | 1885 | 139 | |
| NM_100680.4 | NP_563806.1 | 4 - 5 | 1885 | 139 | ||
| NM_179277.1 | NP_849608.1 | 3 - 4 | 1617 | 139 | ||
Set Membership: |
|