Search
Search

IMPA-4| Gene Symbol: IMPA-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035934.2 | NP_001031011.1 | 2 - 3 | 416 | 81 | |
| NM_100797.6 | NP_172398.1 | 2 - 3 | 503 | 81 | ||
| NM_179292.3 | NP_849623.1 | 2 - 3 | 503 | 81 | ||
| GenBank mRNA | AK316689.1 | - | 2 - 3 | 400 | 81 | |
| AY090944.1 | - | 2 - 3 | 426 | 81 | ||
| AY117313.1 | - | 2 - 3 | 328 | 81 | ||
Set Membership: |
|