Search
Search

NRPB10| Gene Symbol: NRPB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179309.3 | NP_849640.1 | 3 - 4 | 318 | 102 | |
| GenBank mRNA | AK118363.1 | - | 3 - 4 | 280 | 102 | |
| AK228397.1 | - | 3 - 4 | 248 | 102 | ||
| AY084808.1 | - | 3 - 4 | 318 | 102 | ||
Set Membership: |
|