Search
Search

ATSPO11-1| Gene Symbol: ATSPO11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338034.1 | NP_001326966.1 | 5 - 6 | 421 | 91 | |
| NM_001338035.1 | NP_001326967.1 | 5 - 6 | 407 | 91 | ||
| NM_112156.2 | NP_187923.1 | 5 - 6 | 421 | 91 | ||
| GenBank mRNA | AJ251989.1 | - | 5 - 6 | 460 | 91 | |
| BT028955.1 | - | 5 - 6 | 407 | 91 | ||
Set Membership: |
|