Search
Search

REM1| Gene Symbol: REM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119310.4 | NP_567880.1 | 2 - 3 | 617 | 168 | |
| GenBank mRNA | BT003891.1 | - | 2 - 3 | 617 | 168 | |
| BT005002.1 | - | 2 - 3 | 416 | 168 | ||
Set Membership: |
|