Search
Search

HSL1| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342124.1 | NP_001329752.1 | 11 - 12 | 1998 | 68 | |
| NM_119353.4 | NP_194929.2 | 13 - 14 | 2358 | 68 | ||
| GenBank mRNA | AB206554.1 | - | 9 - 10 | 1544 | 68 | |
| AK228013.1 | - | 13 - 14 | 2201 | 68 | ||
Set Membership: |
|