Search
Search

AT4G32050| Gene Symbol: AT4G32050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119357.4 | NP_194933.2 | 8 - 9 | 1191 | 131 | |
| GenBank mRNA | BT021109.1 | - | 8 - 9 | 1104 | 131 | |
Set Membership: |
|