Search
Search

PANK2| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036691.1 | NP_001031768.1 | 3 - 4 | 711 | 111 | |
| NM_001125620.1 | NP_001119092.1 | 3 - 4 | 711 | 111 | ||
| NM_119370.5 | NP_194945.3 | 3 - 4 | 761 | 111 | ||
Set Membership: |
|