Search
Search

AT4G34270| Gene Symbol: AT4G34270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119592.5 | NP_195153.2 | 7 - 8 | 911 | 80 | |
| GenBank mRNA | AY074349.1 | - | 7 - 8 | 814 | 80 | |
| AY091389.1 | - | 7 - 8 | 761 | 80 | ||
Set Membership: |
|