Search
Search

THY-2| Gene Symbol: THY-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342289.1 | NP_001328947.1 | 3 - 4 | 211 | 100 | |
| NM_001342290.1 | NP_001328948.1 | 2 - 3 | 169 | 100 | ||
| NM_119623.5 | NP_195183.2 | 3 - 4 | 211 | 100 | ||
| GenBank mRNA | AK227728.1 | - | 3 - 4 | 150 | 100 | |
Set Membership: |
|