Search
Search

ACR3| Gene Symbol: ACR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036212.1 | NP_001031289.1 | 2 - 3 | 334 | 113 | |
| NM_001124136.2 | NP_001117608.1 | 2 - 3 | 997 | 113 | ||
| NM_106350.3 | NP_565146.1 | 2 - 3 | 377 | 113 | ||
| NM_179565.2 | NP_849896.1 | 2 - 3 | 488 | 113 | ||
| NM_179566.2 | NP_849897.1 | 1 - 2 | 584 | 113 | ||
| GenBank mRNA | AF360322.1 | - | 2 - 3 | 308 | 113 | |
| AF528059.1 | - | 2 - 3 | 257 | 113 | ||
| AK317369.1 | - | 1 - 2 | 567 | 113 | ||
| AY113891.1 | - | 1 - 2 | 65 | 113 | ||
Set Membership: |
|