Search
Search

AT2G19160| Gene Symbol: AT2G19160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335624.1 | NP_001325211.1 | 9 - 10 | 1582 | 76 | |
| NM_179654.3 | NP_849985.1 | 10 - 11 | 1803 | 76 | ||
| GenBank mRNA | AY062829.1 | - | 10 - 11 | 1520 | 76 | |
| BT015873.1 | - | 9 - 10 | 1054 | 76 | ||
| KJ138909.1 | - | 9 - 10 | 1054 | 76 | ||
Set Membership: |
|