Search
Search

SR1| Gene Symbol: SR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124889.2 | NP_001118361.1 | 8 - 9 | 1035 | 93 | |
| NM_001335795.1 | NP_001325089.1 | 7 - 8 | 1561 | 93 | ||
| NM_179692.3 | NP_850023.1 | 7 - 8 | 1107 | 93 | ||
| GenBank mRNA | AF506697.1 | - | 7 - 8 | 731 | 93 | |
| AY510025.1 | - | 7 - 8 | 731 | 93 | ||
| BT002459.1 | - | 7 - 8 | 914 | 93 | ||
Set Membership: |
|