Search
Search

CIPK3| Gene Symbol: CIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036350.1 | NP_001031427.1 | 2 - 3 | 352 | 132 | |
| NM_001336096.1 | NP_001318295.1 | 2 - 3 | 358 | 132 | ||
| NM_179761.2 | NP_850092.1 | 2 - 3 | 352 | 132 | ||
| NM_179763.4 | NP_850094.1 | 3 - 4 | 457 | 132 | ||
| NM_179764.3 | NP_850095.2 | 3 - 4 | 530 | 132 | ||
| GenBank mRNA | AF367290.1 | - | 2 - 3 | 352 | 132 | |
| AY059163.1 | - | 2 - 3 | 234 | 132 | ||
| AY091098.1 | - | 2 - 3 | 345 | 132 | ||
| AY142671.1 | - | 2 - 3 | 234 | 132 | ||
| AY266298.1 | - | 2 - 3 | 234 | 132 | ||
Set Membership: |
|