Search
Search

MT3| Gene Symbol: MT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202970.1 | NP_001189899.1 | 2 - 3 | 357 | 70 | |
| NM_112401.2 | NP_566509.1 | 2 - 3 | 314 | 70 | ||
| GenBank mRNA | AK229545.1 | - | 2 - 3 | 184 | 70 | |
| AY074340.1 | - | 2 - 3 | 181 | 70 | ||
| AY087213.1 | - | 2 - 3 | 178 | 70 | ||
| AY091386.1 | - | 2 - 3 | 102 | 70 | ||
Set Membership: |
|