Search
Search

FAD5| Gene Symbol: FAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112455.4 | NP_566529.1 | 3 - 4 | 1297 | 83 | |
| GenBank mRNA | AB113831.1 | - | 3 - 4 | 811 | 83 | |
| BT000957.1 | - | 3 - 4 | 811 | 83 | ||
Set Membership: |
|