Search
Search

AHP4| Gene Symbol: AHP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202983.2 | NP_001189912.1 | 3 - 4 | 400 | 85 | |
| NM_112507.1 | NP_566544.1 | 2 - 3 | 238 | 85 | ||
| GenBank mRNA | AB041766.1 | - | 2 - 3 | 238 | 85 | |
Set Membership: |
|