Search
Search

PLDP1| Gene Symbol: PLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338257.1 | NP_001326709.1 | 18 - 19 | 3374 | 85 | |
| NM_112553.4 | NP_188302.2 | 18 - 19 | 3420 | 85 | ||
| GenBank mRNA | AF411833.1 | - | 18 - 19 | 3227 | 85 | |
Set Membership: |
|