Search
Search

HCF107| Gene Symbol: HCF107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202985.2 | NP_001189914.1 | 7 - 8 | 1427 | 95 | |
| NM_001338271.1 | NP_001327904.1 | 5 - 6 | 1814 | 95 | ||
| NM_001338272.1 | NP_001327903.1 | 7 - 8 | 1526 | 95 | ||
| NM_112580.5 | NP_188329.1 | 6 - 7 | 1619 | 95 | ||
| GenBank mRNA | AY093112.1 | - | 6 - 7 | 1492 | 95 | |
| BT008405.1 | - | 6 - 7 | 1444 | 95 | ||
Set Membership: |
|