Search Thermo Fisher Scientific
Gene Symbol: POK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338286.1 | NP_001326050.1 | 21 - 22 | 3802 | 131 | |
NM_001338287.1 | NP_001326049.1 | 21 - 22 | 3761 | 131 | ||
NM_112614.4 | NP_188362.2 | 21 - 22 | 3778 | 131 | ||
GenBank mRNA | DQ399529.1 | - | 21 - 22 | 3550 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |