Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338305.1 | NP_001325958.1 | 3 - 4 | 1060 | 118 | |
NM_001338306.1 | NP_001325957.1 | 3 - 4 | 1060 | 118 | ||
NM_112650.4 | NP_188396.2 | 3 - 4 | 1060 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |