Search
Search

CNBT1| Gene Symbol: CNBT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338307.1 | NP_001326341.1 | 1 - 2 | 657 | 74 | |
| NM_112651.5 | NP_566585.1 | 1 - 2 | 755 | 74 | ||
| GenBank mRNA | AF148541.1 | - | 1 - 2 | 634 | 74 | |
Set Membership: |
|