Search
Search

BGLU43| Gene Symbol: BGLU43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084707.2 | NP_001078176.1 | 5 - 6 | 866 | 76 | |
| NM_001338322.1 | NP_001325994.1 | 8 - 9 | 1106 | 76 | ||
| NM_001338324.1 | NP_001325997.1 | 5 - 6 | 866 | 76 | ||
| NM_001338325.1 | NP_001325995.1 | 4 - 5 | 634 | 76 | ||
| NM_001338326.1 | NP_001325996.1 | 4 - 5 | 634 | 76 | ||
| NM_112689.2 | NP_188435.2 | 8 - 9 | 1097 | 76 | ||
| GenBank mRNA | DQ446670.1 | - | 5 - 6 | 866 | 76 | |
Set Membership: |
|