Search
Search

RACK1C_AT| Gene Symbol: RACK1C_AT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112695.3 | NP_188441.1 | 1 - 2 | 1066 | 69 | |
| GenBank mRNA | AY050338.1 | - | 1 - 2 | 948 | 69 | |
| AY101529.1 | - | 1 - 2 | 886 | 69 | ||
Set Membership: |
|