Search
Search

NUC-L2| Gene Symbol: NUC-L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338347.1 | NP_001325664.1 | 2 - 3 | 203 | 88 | |
| NM_001338348.1 | NP_001325663.1 | 2 - 3 | 146 | 88 | ||
| NM_112747.3 | NP_188491.1 | 2 - 3 | 88 | 88 | ||
| GenBank mRNA | DQ446672.1 | - | 2 - 3 | 79 | 88 | |
Set Membership: |
|