Search
Search

SCO2| Gene Symbol: SCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112809.4 | NP_566627.1 | 1 - 2 | 419 | 78 | |
| GenBank mRNA | AY059925.1 | - | 1 - 2 | 398 | 78 | |
| AY081606.1 | - | 1 - 2 | 367 | 78 | ||
| EU402409.1 | - | 1 - 2 | 367 | 78 | ||
Set Membership: |
|