Search
Search

LCB1| Gene Symbol: LCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036719.2 | NP_001031796.1 | 7 - 8 | 805 | 88 | |
| NM_001342410.1 | NP_001329288.1 | 7 - 8 | 1075 | 88 | ||
| NM_119811.3 | NP_568005.1 | 7 - 8 | 1075 | 88 | ||
| GenBank mRNA | AB063254.1 | - | 6 - 7 | 617 | 88 | |
| AY120759.1 | - | 7 - 8 | 893 | 88 | ||
| BT000131.1 | - | 6 - 7 | 596 | 88 | ||
Set Membership: |
|