Search
Search

LEJ2| Gene Symbol: LEJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119855.4 | NP_195409.1 | 5 - 6 | 721 | 61 | |
| GenBank mRNA | AY085164.1 | - | 5 - 6 | 550 | 61 | |
| BT003835.1 | - | 5 - 6 | 536 | 61 | ||
| BT005190.1 | - | 5 - 6 | 460 | 61 | ||
Set Membership: |
|