Search
Search

SPT| Gene Symbol: SPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119857.3 | NP_568010.1 | 5 - 6 | 1079 | 71 | |
| GenBank mRNA | AF319540.1 | - | 5 - 6 | 1017 | 71 | |
| AK229267.1 | - | 5 - 6 | 1035 | 71 | ||
| BT026462.1 | - | 5 - 6 | 995 | 71 | ||
Set Membership: |
|