Search
Search

CYCD5;1| Gene Symbol: CYCD5;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119926.5 | NP_195478.2 | 4 - 5 | 1115 | 158 | |
| GenBank mRNA | AK221431.1 | - | 4 - 5 | 1004 | 158 | |
| BT011732.1 | - | 4 - 5 | 786 | 158 | ||
Set Membership: |
|