Search
Search

HSP60-2| Gene Symbol: HSP60-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202736.1 | NP_001189665.1 | 14 - 15 | 1594 | 139 | |
| NM_179872.3 | NP_850203.1 | 14 - 15 | 1637 | 139 | ||
| GenBank mRNA | AK175415.1 | - | 14 - 15 | 1596 | 139 | |
| AY136360.1 | - | 14 - 15 | 1473 | 139 | ||
| BT000103.1 | - | 14 - 15 | 1433 | 139 | ||
Set Membership: |
|