Search
Search

YLS5| Gene Symbol: YLS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129443.2 | NP_030434.1 | 3 - 4 | 1015 | 136 | |
| NM_179972.3 | NP_850303.1 | 3 - 4 | 1174 | 136 | ||
| GenBank mRNA | AB047808.1 | - | 3 - 4 | 1021 | 136 | |
| AY048272.1 | - | 3 - 4 | 1038 | 136 | ||
| AY084644.1 | - | 3 - 4 | 1015 | 136 | ||
| AY090381.1 | - | 3 - 4 | 1014 | 136 | ||
Set Membership: |
|