Search
Search

MPK11| Gene Symbol: MPK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123738.2 | NP_001117210.1 | 1 - 2 | 398 | 53 | |
| NM_001331281.1 | NP_001322112.1 | 1 - 2 | 406 | 53 | ||
| NM_001331282.1 | NP_001322113.1 | 1 - 2 | 373 | 53 | ||
| NM_100038.4 | NP_563631.2 | 1 - 2 | 373 | 53 | ||
Set Membership: |
|