Search
Search

PFC1| Gene Symbol: PFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331308.1 | NP_001322242.1 | 1 - 2 | 524 | 100 | |
| NM_001331309.1 | NP_001322241.1 | 1 - 2 | 501 | 100 | ||
| NM_001331310.1 | NP_001322240.1 | 2 - 3 | 414 | 100 | ||
| NM_001331311.1 | NP_001322239.1 | 2 - 3 | 391 | 100 | ||
| NM_100068.3 | NP_171690.1 | 2 - 3 | 414 | 100 | ||
| GenBank mRNA | AF051326.1 | - | 2 - 3 | 379 | 100 | |
| AK117412.1 | - | 2 - 3 | 386 | 100 | ||
| BT005097.1 | - | 2 - 3 | 379 | 100 | ||
Set Membership: |
|