Search
Search

MC1| Gene Symbol: MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100097.5 | NP_171719.2 | 3 - 4 | 1049 | 89 | |
| GenBank mRNA | AY219826.1 | - | 3 - 4 | 830 | 89 | |
| AY322525.1 | - | 3 - 4 | 830 | 89 | ||
Set Membership: |
|