Search
Search

MEA| Gene Symbol: MEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331345.1 | NP_001322965.1 | 5 - 6 | 753 | 79 | |
| NM_100139.4 | NP_563658.1 | 5 - 6 | 753 | 79 | ||
| GenBank mRNA | AF060485.1 | - | 5 - 6 | 703 | 79 | |
Set Membership: |
|