Search
Search

GATL5| Gene Symbol: GATL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100152.3 | NP_171772.1 | 1 - 2 | 1548 | 104 | |
| GenBank mRNA | AK118584.1 | - | 1 - 2 | 1405 | 104 | |
| BT002062.1 | - | 1 - 2 | 1159 | 104 | ||
Set Membership: |
|